missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL14 / BRAK Protein
A DNA sequence encoding the Mouse CXCL14 / BRAK, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10260-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_062514.2 | |
CXCL14 / BRAK Protein | |
Research Use Only | |
Cxcl14 | |
The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 1.0-10 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
57266 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SKCKCSRKGP KIRYSDVKKL EMKPKYPHCE EKMVIVTTKS MSRYRGQEHC LHPKLQSTKR FIKWYNAWNE KRRVYEE | |
Greater than 95% as determined by SDS-PAGE and HPLC. |