missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CXCL12 / SDF-1 Protein

A DNA sequence encoding the Mouse CXCL12 / SDF-1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10258-ec-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913268

  • 114.00€ / 2 microgramos


Fecha estimada de envío
13-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_038683.1
CXCL12 / SDF-1 Protein
2 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK
20315
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
CXCL12
The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 50-100 ng/ml.
Mouse
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos