missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CXCL10 / Crg-2 Protein

Recombinant Protein

Marca:  enQuireBio™ QP5448-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916149

  • 3207.00€ / 1 miligramo


Fecha de envío estimada
del proveedor 10-04-2019
Añadir a la cesta

Especificación

Especificación

NP_067249.1
CXCL10 / Crg-2 Protein
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Mouse CXCL10 / Crg-2 induces Primary human T cell chemotaxis (significanty different from control at Mouse CXCL10 / Crg-2 concentrations greater than 5 ng/ml).
Mouse
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
15945
Greater than 95% as determined by SDS-PAGE.
Research Use Only
CXCL10
Recombinant Protein
E. coli
IPLARTVRCN CIHIDDGPVR MRAIGKLEII PASLSCPRVE IIATMKKNDE QRCLNPESKT IKNLMKAFSQ KRSKRAP
Videos