missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CTGF Protein

A DNA sequence encoding the Mus musculus (Mouse) CTGF, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5885-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15928459

  • 985.00€ / 200 microgramos


Fecha estimada de envío
30-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_034347.2
CTGF Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
14219
Greater than 80% as determined by SDS-PAGE.
Research Use Only
CTGF
Recombinant Protein
E. coli
QDCSAQCQCA AEAAPHCPAG VSLVLDGCGC CRVCAKQLGE LCTERDPCDP HKGLFCDFGS PANRKIGVCT AKDGAPCVFG GSVYRSGESF QSSCKYQCTC LDGAVGCVPL CSMDVRLPSP DCPFPRRVKL PGKCCEEWVC DEPKDRTAVG PALAAYRLED TFGPDPTMMR ANCLVQTTEW SACSKTCGMG ISTRVTNDNT FCRLEKQSRL CMVRPCEADL EENIKKGKKC IRTPKIAKPV KFELSGCTSV KTYRAKFCGV CTDGRCCTPH RTTTLPVEFK CPDGEIMKKN MMFIKTCACH YNCPGDNDIF ESLYYRKMY
Videos
Documentos