missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CNDP1 Protein
A cDNA sequence encoding the CNDP1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11467-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CNDP1 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MFSSAHSGLL EKLFHYIDLH QDEFVQTLKE WVAIESDSVQ PVPRLRQKLF QMMALAADKL RNLGAGVESI DLGSQQMPDG QSLPIPPILL AELGSDPEKP TVCFYGHLDV QPAQKDDGWL TDPYTLTEVD GKLYGRGATD NKGPVLAWIN AVSTFRALQQ DLPVNIKLIL EGMEEAGSIA LEELVMREKD HFFSSVDYIV ISDNLWLSQR KPALTYGTRG NCYFTVEVKC RDQDFHSGTF GGILNEPMAD LVALLGSLVD SSGHILIPGI YDQMAPITEG EKTMYKNIDM DLEEYQNINQ VEKFLFDTKE ELLMHLWRYP SLSIHGIEGA FDEPGTKTVI PGRVLGKFSI RLVPTMSPSV VEKQVTQHLE AVFSKRNSFN KMAVSMVLGL HPWTANVNDT QYLAAQRTIK TVFGVNPDMI RDGSTIPIAK IFQAITQKSV MMLPLGAVDD GEHSQNEKIN RWNYIQGSKL FAAFFLELSK QHSGHQMPSS VYLEHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
CNDP1 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |