missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD93 Protein
A cDNA sequence encoding the CD93 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11356-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
17064 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADSQAVVCEG TACYTAHWGK LSAAEAQHRC NENGGNLATV KSEEEARHVQ QALTQLLKTK APLEAKMGKF WIGLQREKGN CTYHDLPMRG FSWVGGGEDT AYSNWYKASK SSCIFKRCVS LILDLSLTPH PSHLPKWHES PCGTPEAPGN SIEGFLCKFN FKGMCRPLAL GGPGRVTYTT PFQATTSSLE AVPFASVANV ACGDEAKSET HYFLCNEKTP GIFHWGSSGP LCVSPKFGCS FNNGGCQQDC FEGGDGSFRC GCRPGFRLLD DLVTCASRNP CSSNPCTGGG MCHSVPLSEN YTCRCPSGYQ LDSSQVHCVD IDECQDSPCA QDCVNTLGSF HCECWVGYQP SGPKEEACED VDECAAANSP CAQGCINTDG SFYCSCKEGY IVSGEDSTQC EDIDECSDAR GNPCDSLCFN TDGSFRCGCP PGWELAPNGV FCSRGTVFSE LPARPPQKED NDDRKESTMP PTEMPSSPSG SKDVSNRAQT TGLFVQSDIP TASVPLEIEI PSEVSDVWFE LGTYLPTTSG HSKPTHEDSV SAHSDTDGQN LEHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
CD93 Protein | |
Research Use Only | |
Cd93 | |
Mouse | |
His | |
CD93 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |