missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD90 / THY-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) CD90 / THY-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6789-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_033408.1 | |
CD90 / THY-1 Protein | |
Research Use Only | |
Thy1 | |
Recombinant Protein | |
E. coli | |
QKVTSLTACL VNQNLRLDCR HENNTKDNSI QHEFSLTREK RKHVLSGTLG IPEHTYRSRV TLSNQPYIKV LTLANFTTKD EGDYFCELQV SGANPMSSNK SISVYRDKLV KC | |
Greater than 90% as determined by SDS-PAGE. |
21838 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |