missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD81 / TAPA-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) CD81 / TAPA-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5798-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_598416.1 | |
CD81 / TAPA-1 Protein | |
Research Use Only | |
Cd81 | |
Recombinant Protein | |
E. coli | |
KDQIAKDVKQ FYDQALQQAV MDDDANNAKA VVKTFHETLN CCGSNALTTL TTTILRNSLC PSGGNILTPL LQQDCHQKID ELFSGK | |
Greater than 90% as determined by SDS-PAGE. |
12520 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |