missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD36 Protein

A cDNA sequence encoding the CD36 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11329-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916888

  • 5941.00€ / 1 miligramo


Fecha estimada de envío
17-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

12491
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Cd36
Mouse
His
CD36 protein solution (1 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol.
CD36 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Insect
ADPGDMLIEK TIKREVVLEE GTTAFKNWVK TGTTVYRQFW IFDVQNPDDV AKNSSKIKVK QRGPYTYRVR YLAKENITQD PEDHTVSFVQ PNGAIFEPSL SVGTEDDNFT VLNLAVAAAP HIYQNSFVQV VLNSLIKKSK SSMFQTRSLK ELLWGYKDPF LSLVPYPIST TVGVFYPYND TVDGVYKVFN GKDNISKVAI IESYKGKRNL SYWPSYCDMI NGTDAASFPP FVEKSRTLRF FSSDICRSIY AVFGSEIDLK GIPVYRFVLP ANAFASPLQN PDNHCFCTEK VISNNCTSYG VLDIGKCKEG KPVYISLPHF LHASPDVSEP IEGLHPNEDE HRTYLDVEPI TGFTLQFAKR LQVNILVKPA RKIEALKNLK RPYIVPILWL NETGTIGDEK AEMFKTQVTG KIKHHHHHH
Videos