missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD274 Protein
A cDNA sequence encoding the CD274 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11320-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
60533 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMFTITA PKDLYVVEYG SNVTMECRFP VERELDLLAL VVYWEKEDEQ VIQFVAGEED LKPQHSNFRG RASLPKDQLL KGNAALQITD VKLQDAGVYC CIISYGGADY KRITLKVNAP YRKINQRISV DPATSEHELI CQAEGYPEAE VIWTNSDHQPVSGKRSVTTS RTEGMLLNVT SSLRVNATAN DVFYCTFWRS QPGQNHTAEL IIPELPATHP PQNRTH | |
Greater than 85% as determined by SDS-PAGE. |
CD274 Protein | |
Research Use Only | |
Cd274 | |
Mouse | |
His | |
CD274 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.15M NaCl. |