missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CCL4 / MIP1B Protein
Recombinant Protein
Marca: enQuireBio™ QP5452-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_038680.1 | |
CCL4 / MIP1B Protein | |
Research Use Only | |
CCL4 | |
Recombinant Protein | |
E. coli | |
APMGSDPPTS CCFSYTSRQL HRSFVMDYYE TSSLCSKPAV VFLTKRGRQI CANPSEPWVT EYMSDLELN | |
Greater than 95% as determined by SDS-PAGE. |
20303 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Appropriate THP-1 chemotaxis is confirmed for each lot of Mouse CCL4 / MIP1B. To pass QC, Mouse CCL4 / MIP1B must significantly induced cell migration at concentrations below 100 ng/ml. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |