missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CCL12 / MCP-5 Protein
A DNA sequence encoding the Mouse CCL12 / MCP-5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10271-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_035461.2 | |
CCL12 / MCP-5 Protein | |
Research Use Only | |
Ccl12 | |
The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 10-50 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
20293 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GPDAVSTPVT CCYNVVKQKI HVRKLKSYRR ITSSQCPREA VIFRTILDKE ICADPKEKWV KNSINHLDKT SQTFILEPSC LG | |
Greater than 95% as determined by SDS-PAGE and HPLC. |