missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Cathepsin G Protein

A DNA sequence encoding the Mus musculus (Mouse) Cathepsin G, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5887-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15928469

  • 245.00€ / 10 microgramos


Fecha estimada de envío
25-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_031826.1
Cathepsin G Protein
10 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
13035
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Ctsg
Recombinant Protein
E. coli
IIGGREARPH SYPYMAFLLI QSPEGLSACG GFLVREDFVL TAAHCLGSSI NVTLGAHNIQ MRERTQQLIT VLRAIRHPDY NPQNIRNDIM LLQLRRRARR SGSVKPVALP QASKKLQPGD LCTVAGWGRV SQSRGTNVLQ EVQLRVQMDQ MCANRFQFYN SQTQICVGNP RERKSAFRGD SGGPLVCSNV AQGIVSYGSN NGNPPAVFTK IQSFMPWIKR TMRRFAPRYQ RPANSLSQAQ T
Videos
Documentos