missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CALR / Calreticulin Protein

A DNA sequence encoding the Mus musculus (Mouse) CALR / Calreticulin, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5750-ye-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15937499

  • 1705.00€ / 500 microgramos


Fecha estimada de envío
26-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_031617.1
CALR / Calreticulin Protein
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
12317
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Calr
Recombinant Protein
Yeast
DPAIYFKEQF LDGDAWTNRW VESKHKSDFG KFVLSSGKFY GDLEKDKGLQ TSQDARFYAL SAKFEPFSNK GQTLVVQFTV KHEQNIDCGG GYVKLFPSGL DQKDMHGDSE YNIMFGPDIC GPGTKKVHVI FNYKGKNVLI NKDIRCKDDE FTHLYTLIVR PDNTYEVKID NSQVESGSLE DDWDFLPPKK IKDPDAAKPE DWDERAKIDD PTDSKPEDWD KPEHIPDPDA KKPEDWDEEM DGEWEPPVIQ NPEYKGEWKP RQIDNPDYKG TWIHPEIDNP EYSPDANIYA YDSFAVLGLD LWQVKSGTIF DNFLITNDEA YAEEFGNETW GVTKAAEKQM KDKQDEEQRL KEEEEDKKRK EEEEAEDKED DDDRDEDEDE EDEKEEDEEE SPGQAKDEL
Videos
Documentos