missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse C-C motif chemokine 21c Protein

A DNA sequence encoding the Mouse C-C motif chemokine 21c, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10275-ec-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913338

  • 105.00€ / 5 microgramos


Fecha estimada de envío
25-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001180595.1
C-C motif chemokine 21c Protein
5 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
SDGGGQDCCL KYSQKKIPYS IVRGYRKQEP SLGCPIPAIL FLPRKHSKPE LCANPEEGWV QNLMRRLDQP PAPGKQSPGC RKNRGTSKSG KKGKGSKGCK RTEQTQPSRG
100041504
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
Ccl21c
The biological activity determined by a chemotaxis bioassay using murine T-lymphocytes is in a concentration range of 10-100 ng/ml.
Mouse
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos