missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse bFGF / FGF2 Protein

A DNA sequence encoding the Mus musculus (Mouse) bFGF / FGF2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8538-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055171

  • 1715.00€ / 500 microgramos


Fecha estimada de envío
20-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_032032.1
bFGF / FGF2 Protein
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
14173
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FGF2
Recombinant Protein
E. coli
PALPEDGGAA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHV KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTEE CFFFERLESN NYNTYRSRKY SSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
Videos
Documentos