missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Arginase-2, mitochondrial Protein
A DNA sequence encoding the Mus musculus (Mouse) Arginase-2, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5677-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_033835.1 | |
Arginase-2, mitochondrial Protein | |
Research Use Only | |
Arg2 | |
Recombinant Protein | |
E. coli | |
VHSVAIVGAP FSRGQKKLGV EYGPAAIREA GLLKRLSRLG CHLKDFGDLS FTNVPQDDPY NNLVVYPRSV GLANQELAEV VSRAVSGGYS CVTMGGDHSL AIGTIIGHAR HRPDLCVIWV DAHADINTPL TTVSGNIHGQ PLSFLIKELQ DKVPQLPGFS WIKPCLSPPN IVYIGLRDVE PPEHFILKNY DIQYFSMREI DRLGIQKVME QTFDRLIGKR QRPIHLSFDI DAFDPKLAPA TGTPVVGGLT YREGVYITEE IHNTGLLSAL DLVEVNPHLA TSEEEAKATA RLAVDVIASS FGQTREGGHI VYDHLPTPSS PHESENEECV RI | |
Greater than 90% as determined by SDS-PAGE. |
11847 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |