missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Apolipoprotein A-IV Protein

A DNA sequence encoding the Mus musculus (Mouse) Apolipoprotein A-IV, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5662-ye-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15936819

  • 342.00€ / 50 microgramos


Fecha estimada de envío
27-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_031494.2
Apolipoprotein A-IV Protein
50 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
11808
Greater than 90% as determined by SDS-PAGE.
Research Use Only
APOA4
Recombinant Protein
Yeast
EVTSDQVANV VWDYFTQLSN NAKEAVEQFQ KTDVTQQLST LFQDKLGDAS TYADGVHNKL VPFVVQLSGH LAQETERVKE EIKKELEDLR DRMMPHANKV TQTFGENMQK LQEHLKPYAV DLQDQINTQT QEMKLQLTPY IQRMQTTIKE NVDNLHTSMM PLATNLKDKF NRNMEELKGH LTPRANELKA TIDQNLEDLR RSLAPLTVGV QEKLNHQMEG LAFQMKKNAE ELQTKVSAKI DQLQKNLAPL VEDVQSKVKG NTEGLQKSLE DLNRQLEQQV EEFRRTVEPM GEMFNKALVQ QLEQFRQQLG PNSGEVESHL SFLEKSLREK VNSFMSTLEK KGSPDQPQAL PLPEQAQEQA QEQAQEQVQP KPLES
Videos
Documentos