missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse AIMP1 / EMAP2 / SCYE1 Protein
A DNA sequence encoding the Mus musculus (Mouse) AIMP1 / EMAP2 / SCYE1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6654-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
AIMP1 / EMAP2 / SCYE1 Protein | |
Research Use Only | |
Aimp1 | |
Recombinant Protein | |
E. coli | |
ATNDAVLKRL EQKGAEADQI IEYLKQQVAL LKEKAILQAT MREEKKLRVE NAKLKKEIEE LKQELILAEI HNGVEQVRVR LSTPLQTNCT ASESVVQSPS VATTASPATK EQIKAGEEKK VKEKTEKKGE KKEKQQSAAA STDSKPIDAS RLDLRIGCIV TAKKHPDADS LYVEEVDVGE AAPRTVVSGL VNHVPLEQMQ NRMVVLLCNL KPAKMRGVLS QAMVMCASSP EKVEILAPPN GSVPGDRITF DAFPGEPDKE LNPKKKIWEQ IQPDLHTNAE CVATYKGAPF EVKGKGVCRA QTMANSGIK | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |