missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse AHCY Protein
A cDNA sequence encoding the AHCY was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10981-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
11615; 269378 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSDKLP YKVADIGLAA WGRKALDIAE NEMPGLMRMR EMYSASKPLK GARIAGCLHM TVETAVLIET LVALGAEVRW SSCNIFSTQD HAAAAIAKAG IPVFAWKGET DEEYLWCIEQ TLHFKDGPLN MILDDGGDLT NLIHTKYPQL LSGIRGISEE TTTGVHNLYK MMSNGILKVP AINVNDSVTK SKFDNLYGCR ESLIDGIKRA TDVMIAGKVA VVAGYGDVGK GCAQALRGFG ARVIITEIDP INALQAAMEG YEVTTMDEAC KEGNIFVTTT GCVDIILGRH FEQMKDDAIV CNIGHFDVEI DVKWLNENAV EKVNIKPQVD RYWLKNGRRI ILLAEGRLVN LGCAMGHPSF VMSNSFTNQV MAQIELWTHP DKYPVGVHFL PKKLDEAVAE AHLGKLNVKL TKLTEKQAQY LGMPINGPFK PDHYRY | |
Greater than 90.0% as determined by SDS-PAGE. |
AHCY Protein | |
Research Use Only | |
Ahcy | |
Mouse | |
His | |
AHCY protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) containing 40% glycerol 0.2M NaCl, 1mM DTT. |