missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Adamts5 Protein

A DNA sequence encoding the Mus musculus (Mouse) Adamts5, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8228-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053411

  • 437.00€ / 50 microgramos


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_035912.2
Adamts5 Protein
50 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
23794
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Adamts5
Recombinant Protein
E. coli
SISRARQVEL LLVADSSMAR MYGRGLQHYL LTLASIANRL YSHASIENHI RLAVVKVVVL TDKDTSLEVS KNAATTLKNF CKWQHQHNQL GDDHEEHYDA AILFTREDLC GHHSCDTLGM ADVGTICSPE RSCAVIEDDG LHAAFTVAHE IGHLLGLSHD DSKFCEENFG TTEDKRLMSS ILTSIDASKP WSKCTSATIT EFLDDGHGNC LLDLPRKQIL GPEELPGQTY DATQQCNLTF GPEYSVCPGM DVCARLWCAV VRQGQMVCLT KKLPAVEGTP CGKGRVCLQG KCVDKTKKKY YSTSSHGNWG SWGPWGQCSR SCGGGVQFAY RHCNNPAPRN SGRYCTGKRA IYRSCSVTPC PPNGKSFRHE QCEAKNGYQS DAKGVKTFVE WVPKYAGVLP ADVCKLTCRA KGTGYYVVFS PKVTDGTECR PYSNSVCVRG RCVRTGCDGI IGSKLQYDKC GVCGGDNSSC TKIIGTFNKK SKGYTDVVRI PEGATHIKVR QFKAKDQTRF TAYLALKKKT GEYLINGKYM ISTSETIIDI NGTVMNYSGW SHRDDFLHGM GYSATKEILI VQILATDPTK ALDVRYSFFV PKKTTQKVNS VISHGSNKVG PHSTQLQWVT GPWLACSRTC DTGWHTRTVQ CQDGNRKLAK GCLLSQRPSA FKQCLLKKC
Videos
Documentos