missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Actin, cytoplasmic 1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Actin, cytoplasmic 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8585-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055421

  • 2826.00€ / 1 miligramo


Fecha estimada de envío
23-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_031419.1
Actin, cytoplasmic 1 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Mouse
GST
Tris-based buffer, 50% glycerol
11461
Greater than 90% as determined by SDS-PAGE.
Research Use Only
actin
Recombinant Protein
E. coli
DNGSGMCKAG FAGDDAPRAV FPSIVGRPRH QGVMVGMGQK DSYVGDEAQS KRGILTLKYP IEHGIVTNWD DMEKIWHHTF YNELRVAPEE HPVLLTEAPL NPKANREKMT QIMFETFNTP AMYVAIQAVL SLYASGRTTG IVMDSGDGVT HTVPIYEGYA LPHAILRLDL AGRDLTDYLM KILTERGYSF TTTAEREIVR DIKEKLCYVA LDFEQEMATA ASSSSLEKSY ELPDGQVITI GNERFRCPEA LFQPSFLGME SCGIHETTFN SIMKCDVDIR KDLYANTVLS GGTTMYPGIA DRMQKEITAL APSTMKIKII APPERKYSVW IGGSILASLS TFQQMWISKQ EYDESGPSIV HRKCF
Videos
Documentos