missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse 4-1BBR Protein
A cDNA sequence encoding the 4-1BBR was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10920-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
4-1BBR Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
VQNSCDNCQP GTFCRKYNPV CKSCPPSTFS SIGGQPNCNI CRVCAGYFRF KKFCSSTHNA ECECIEGFHC LGPQCTRCEK DCRPGQELTK QGCKTCSLGT FNDQNGTGVC RPWTNCSLDG RSVLKTGTTE KDVVCGPPVV SFSPSTTISV TPEGGPGGHS LQVLLEHHHH HH | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
4-1BBR protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |