missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Monkey GM CSF Protein

A cDNA sequence encoding the GM CSF was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10647-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914518

  • 103.00€ / 2 microgramos


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

Monkey GM CSF Protein
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
APARSPSPGT QPWEHVNAIQ EARRLLNLSR DTAAEMNKTV EVVSEMFDLQ EPSCLQTRLE LYKQGLQGSL TKLKGPLTMM ASHYKQHCPP TPETSCATQI ITFQSFKENL KDFLLVIPFD CWEPVQE
Greater than 98.0% as determined by SDS-PAGE and RP-HPLC.
Research Use Only
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 0.1ng/ml, corresponding to a specific activity of > 10,000,000 IU/mg.
Donkey
Untagged
GM-CSF was lyophilized from a concentrated (1 mg/ml) solution containing 1x PBS pH 7.4.
Videos