missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque Serum Albumin / HSA / ALB Protein

A DNA sequence encoding the Macaca fascicularis (Crab-eating macaque) (Cynomolgus monkey) Serum Albumin / HSA / ALB, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5643-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15936659

  • 1715.00€ / 500 microgramos


Fecha estimada de envío
24-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001306284.1
Serum Albumin / HSA / ALB Protein
500 μg
Not determined.
Full Length
Macaque
His-SUMO
Tris-based buffer, 50% glycerol
102130757
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ALB
Recombinant Protein
E. coli
DTHKSEVAHR FKDLGEEHFK GLVLVAFSQY LQQCPFEEHV KLVNEVTEFA KTCVADESAE NCDKSLHTLF GDKLCTVATL RETYGEMADC CAKQEPERNE CFLQHKDDNP NLPPLVRPEV DVMCTAFHDN EATFLKKYLY EVARRHPYFY APELLFFAAR YKAAFAECCQ AADKAACLLP KLDELRDEGK ASSAKQRLKC ASLQKFGDRA FKAWAVARLS QKFPKAEFAE VSKLVTDLTK VHTECCHGDL LECADDRADL AKYMCENQDS ISSKLKECCD KPLLEKSHCL AEVENDEMPA DLPSLAADYV ESKDVCKNYA EAKDVFLGMF LYEYARRHPD YSVMLLLRLA KAYEATLEKC CAAADPHECY AKVFDEFQPL VEEPQNLVKQ NCELFEQLGE YKFQNALLVR YTKKVPQVST PTLVEVSRNL GKVGAKCCKL PEAKRMPCAE DYLSVVLNRL CVLHEKTPVS EKVTKCCTES LVNRRPCFSA LELDEAYVPK AFNAETFTFH ADMCTLSEKE KQVKKQTALV ELVKHKPKAT KEQLKGVMDN FAAFVEKCCK ADDKEACFAE EGPKFVAASQ AALA
Videos
Documentos