missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque MAPT / Tau Protein
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) MAPT / Tau, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6344-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
XP_014975440.1 | |
MAPT / Tau Protein | |
Research Use Only | |
Mapt | |
Recombinant Protein | |
E. coli | |
AEPRQEFDVM EDHAGTYGLG DRKDQEGYTM LQDQEGDTDA GLKESPLQTP AEDGSEELGS ETSDAKSTPT AEDVTAPLVD ERAPGEQAAA QPHMEIPEGT TAEEAGIGDT PSLEDEAAGH VTQARMVSKS KDGTGSDDKK AKGADGKTKI ATPRGAAPPG QKGQANATRI PAKTPPAPKT PPSSATKQVQ RKPPPAEPTS ERGEPPKSGD RSGYSSPGSP GTPGSRSRTP SLPTPPAREP KKVAVVRTPP KSPSSAKSRL QTAPVPMPDL KNVKSKIGST ENLKHQPGGG KVQIINKKLD LSNVQSKCGS KDNIKHVPGG GSVQIVYKPV DLSKVTSKCG SLGNIHHKPG GGQVEVKSEK LDFKDRVQSK IGSLDNITHV PGGGNKKIET HKLTFRENAK AKTDHGAEIV YKSPVVSGDT SPRHLSNVSS TGSIDMVDSP QLATLADEVS ASLAKQGL | |
Greater than 90% as determined by SDS-PAGE. |
574327 | |
200 μg | |
Not determined. | |
Full Length | |
Macaque | |
His-SUMO | |
Tris-based buffer, 50% glycerol |