missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque IL6 / Interleukin-6 Protein

A DNA sequence encoding the Rhesus Macaque IL6 / Interleukin-6 Protein, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP1556-ec-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15925529

  • 114.00€ / 2 microgramos


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

IL6 / Interleukin-6 Protein
2 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MAPVLPGEDS KNVAAPHSQP LTSSERIDKH IRYILDGISA LRKETCNRSN MCESSKEALA ENNLNLPKMA EKDGCFQSGF NEDTCLVKII TGLLEFEVYL EYLQNRFESS EEQARAVQMS TKVLIQFLQK KAKNLDAITT PEPTTNASLL TKLQAQNQWL QDMTTHLILR SFKEFLQSNL RALRQM
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
IL6
The ED50 as determined by a cell proliferation assay usingIL-6-dependent murine 7TD1 cells is less than 0.1 ng/ml, corresponding to a specific activity of > 1.0 M 107 IU/mg.
Macaque
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos