missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL-3 / Interleukin-3 Protein
A DNA sequence encoding the Rhesus Macaque IL-3 / Interleukin-3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10336-ec-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001095204.1 | |
IL-3 / Interleukin-3 Protein | |
Research Use Only | |
IL3 | |
Assay 1: The ED50 as determined by the dose- dependent stimulation of the proliferation of murine NFS-60 cells is less than 5.0 ng/ml, corresponding to a specific activity of > 2 M 105 IU/mg. Assay 2: The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 0.05 ng/ml, corresponding to a specific activity of > 2.0 M 107 IU/mg. | |
Macaque | |
Untagged | |
0.2 um filtered PBS, pH 7.4, 5 % trehalose, lyophilized |
706946 | |
2 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
APMTQTTSLK TSWAKCSNMI DEIITHLNQP PLPSPDFNNL NEEDQTILVE KNLRRSNLEA FSKAVKSLQN ASAIESILKN LPPCLPMATA APTRPPIRIT NGDRNDFRRK LKFYLKTLEN EQAQ | |
Greater than 98% as determined by SDS-PAGE and HPLC. |