missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque IL-3 / Interleukin-3 Protein

A DNA sequence encoding the Rhesus Macaque IL-3 / Interleukin-3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10336-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913548

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Especificaciones

Especificaciones

NP_001095204.1
IL-3 / Interleukin-3 Protein
Research Use Only
IL3
Assay 1: The ED50 as determined by the dose- dependent stimulation of the proliferation of murine NFS-60 cells is less than 5.0 ng/ml, corresponding to a specific activity of > 2 M 105 IU/mg. Assay 2: The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 0.05 ng/ml, corresponding to a specific activity of > 2.0 M 107 IU/mg.
Macaque
Untagged
0.2 um filtered PBS, pH 7.4, 5 % trehalose, lyophilized
706946
10 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
APMTQTTSLK TSWAKCSNMI DEIITHLNQP PLPSPDFNNL NEEDQTILVE KNLRRSNLEA FSKAVKSLQN ASAIESILKN LPPCLPMATA APTRPPIRIT NGDRNDFRRK LKFYLKTLEN EQAQ
Greater than 98% as determined by SDS-PAGE and HPLC.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales