missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque G-CSF / CSF3 Protein
A DNA sequence encoding the Rhesus Macaque G-CSF / CSF3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10444-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
G-CSF / CSF3 Protein | |
Research Use Only | |
CSF3 | |
The ED50 as determined by a cell proliferation assay using murine NFS-60 cells is less than 0.05 ng/ml, corresponding to a specific activity of > 2.0 M 107 IU/mg. | |
Macaque | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
TPLGPASSLP QSFLLKCLEQ VRKIQGDGAA LQEKLCATYK LCHPEELVLL RHSLGIPWAP LSSCPSQALQ LTGCLSQLHS SLFLYQGLLQ ALEGISPELS PTLDTLQLDI ADFATTIWQQ MEDLGMAPAL QPTQGAMPAF TSAFQRRAGG VLVASHLQRF LELAYRVLRH LAQS | |
Greater than 98% as determined by SDS-PAGE and HPLC. |