missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque Erythropoietin / EPO Protein

A DNA sequence encoding the Macaca mulatta (Rhesus macaque) Erythropoietin / EPO, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10075-ma-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15922828

Cantidad Precio
1 1070.00€ / 50 microgramos
4 998.95€ / 50 microgramos
Fecha estimada de envío
02-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_001036201.1
Erythropoietin / EPO Protein
50 μg
Not determined.
Full Length
Macaque
His
Tris-based buffer, 50% glycerol
719294
Greater than 90% as determined by SDS-PAGE.
Research Use Only
EPO
Recombinant Protein
Mammal
APPRLVCDSR VLERYLLEAK EAENVTMGCS ESCSLNENIT VPDTKVNFYA WKRIEVGQQA VEVWQGLALL SEAVLRGQAV LANSSQPFEP LQLHMDKAIS GLRSITTLLR ALGAQEAISL PDAASAAPLR TITADTFCKL FRVYSNFLRG KLKLYTGEAC RRGDR
Videos
Documentos