missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque CXCL10 / Crg-2 Protein

A DNA sequence encoding the Rhesus Macaque CXCL10 / Crg-2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10245-ec-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933218

  • 105.00€ / 5 microgramos


Fecha estimada de envío
06-08-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001028064.1
CXCL10 / Crg-2 Protein
5 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
IPLSRTVRCT CISISNQPVN PRSLEKLEII PPSQFCPHVE IIATMKKKGE KRCLNPESKA IKNLLKAVSK ERSKRSP
574243
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
CXCL10
The biological activity determined by a chemotaxis bioassay using human peripheral blood T-lymphocytes is in a concentration range of 10- 50 ng/ml.
Macaque
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos