missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque CD40L / CD154 / TNFSF5 Protein

A DNA sequence encoding the Macaca mulatta (Rhesus macaque) CD40L / CD154 / TNFSF5, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5786-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15907749

  • 1857.00€ / 1 miligramo


Fecha estimada de envío
12-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001028011.1
CD40L / CD154 / TNFSF5 Protein
1 mg
Not determined.
Extracellular Domain
Macaque
His
Tris-based buffer, 50% glycerol
574160
Greater than 80% as determined by SDS-PAGE.
Research Use Only
CD40LG
Recombinant Protein
E. coli
VYLHRRLDKI EDERNLHEDF VFMKTIQRCN TGERSLSLLN CEEIKSQFEG FVKDIMLNKE EKKKENSFEM QKGDQNPQIA AHVISEASSK TTSVLQWAEK GYYTMSNNLV TLENGKQLTV KRQGLYYIYA QVTFCSNREA SSQAPFIASL CLKSPGRFER ILLRAANTHS SAKPCGQQSI HLGGVFELQP GASVFVNVTD PSQVSHGTGF TSFGLLKL
Videos