missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque Amyloid protein 1

A DNA sequence encoding the Rhesus Macaque Amyloid protein A, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10247-ec-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913228

  • 114.00€ / 2 microgramos


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

Amyloid protein 1
2 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
RSWFSFLGEA YDGARDMWRA YSDMKEANYK NSDKYFHARG NYDAAQRGPG GVWAAEVISD ARENIQKLLG RGAEDTLADQ AANEWGRSGK DPNHFRPAGL PEKY
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
SAA1
The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 10-100 ng/ml.
Macaque
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos