missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant M. tuberculosis Thymidylate kinase Protein
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Thymidylate kinase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5956-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
WP_003417036.1 | |
10 μg | |
Not determined. | |
Full Length | |
M. tuberculosis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
M. tuberculosis Thymidylate kinase Protein | |
Research Use Only | |
tmk | |
Recombinant Protein | |
E. coli | |
MLIAIEGVDG AGKRTLVEKL SGAFRAAGRS VATLAFPRYG QSVAADIAAE ALHGEHGDLA SSVYAMATLF ALDRAGAVHT IQGLCRGYDV VILDRYVASN AAYSAARLHE NAAGKAAAWV QRIEFARLGL PKPDWQVLLA VSAELAGERS RGRAQRDPGR ARDNYERDAE LQQRTGAVYA ELAAQGWGGR WLVVGADVDP GRLAATLAPP DVPS | |
Greater than 90% as determined by SDS-PAGE. |