missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza Parainfluenza Type-1 Protein

Recombinant Protein

Marca:  enQuireBio™ QP12959-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15912079

  • 103.00€ / 10 microgramos


Fecha estimada de envío
24-06-2019
Añadir a la cesta

Especificaciones

Especificaciones

Influenza Parainfluenza Type-1 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined byAnalysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
The Parainfluenza type-1solution contains STE, 0.02 % sodium azide (NaN3).
Videos