missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza H3N2 Wyoming Protein

A cDNA sequence encoding the H3N2 Wyoming was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12092-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919348

  • 103.00€ / 2 microgramos


Fecha estimada de envío
17-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

Influenza H3N2 Wyoming Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
The Recombinant H3N2 A/Wyoming/2003/3 solution contains 10mM Sodium phosphate, pH 7.4, 150mM NaCl and 0.005% Tween-20.
Videos