missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza H3N2 Shandong Protein

Recombinant Protein

Marca:  enQuireBio™ QP12086-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919328

  • 160.00€ / 50 microgramos


Fecha estimada de envío
22-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

Influenza H3N2 Shandong Protein
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined byAnalysis by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
The H3N2A/ Shandong/9/93 solution (1.0 mg/ml) contains STE, 0.1% sodium azide (NaN3) and 0.005% thimerosal.
Videos