missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza H1N1 Puerto Rico Recombinant Protein

A cDNA sequence encoding the H1N1 Puerto Rico Recombinant was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12077-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919298

  • 230.00€ / 10 microgramos


Fecha de envío estimada
del proveedor 03-04-2019
Añadir a la cesta

Especificación

Especificación

Influenza H1N1 Puerto Rico Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
The Recombinant H1N1 A/Puerto Rico 08/1934 solution contains 10mM Sodium phosphate, pH 7.2, 150mM NaCl and 0.005% Tween-20.
Videos