missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H1N1 Beijing Protein
Recombinant Protein
Marca: enQuireBio™ QP12074-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Influenza H1N1 Beijing Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The H1N1 A/Beijing/262/95 solution contains STE, 0.09% sodium azide (NaN3) and 0.005% thimerosal. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |