missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza-B Tokio Protein

A cDNA sequence encoding the Influenza-B Tokio was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12443-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15910439

  • 103.00€ / 10 microgramos


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

Influenza-B Tokio Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined byAnalysis by SDS-PAGE.
Research Use Only
Recombinant Protein
Allantonic Fluid
The B/Tokio/53/99solution (1.7 mg/ml) contains STE, 0.1% sodium azide (NaN3) and 0.005% thimerosal.
Videos