missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza-B Malaysia Protein
A cDNA sequence encoding the Influenza-B Malaysia was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12440-EGG-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Influenza-B Malaysia Protein | |
Research Use Only | |
Recombinant Protein | |
Influenza B virus | |
The Recombinant B/Malaysia/2506/2004 solution contains 10mM Sodium Phosphate, pH 7.0 and 150mM NaCl, 0.005% Tween-20. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |