Learn More
enQuireBio™ Recombinant Influenza A H2N2 (Mallard / New York / 6750 / 1978) Hemagglutinin Protein
A DNA sequence encoding the Influenza A virus (strain A/Mallard/New York/6750/1978 H2N2) Influenza A H2N2 (strain A/Mallard/New York/6750/1978) Hemagglutinin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7702-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Influenza A H2N2 (Mallard / New York / 6750 / 1978) Hemagglutinin Protein | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
HA | |
Recombinant Protein | |
E. coli | |
DQICIGYHAN NSTEKVDTIL ERNVTVTHAK DILEKTHNGK LCRLSGIPPL ELGDCSIAGW LLGNPECDRL LSVPEWSYIV EKENPANGLC YPGNFNDYEE LKHLLTRVTH FEKIKILPRD QWTQHTTTGG SRACAVSGNP SFFRNMVWLT KKGSNYPVAK GSYNNTSGEQ MLVIWGIHHP NDDTEQRTLY QNVGTYVSVG TSTLNKRSIP EIATRPKVNG QGGRMEFSWT LLETWDVINF ESTGNLIAPE YGFKISKRGS SGIMKTEKTL ENCETKCQTP LGAINTTLPF HNIHPLTIGE CPKYVKSDRL VLATGLRNVP QIES |