missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zyxin Protein
A DNA sequence encoding the Homo sapiens (Human) Zyxin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10108-ma-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001010972.1 | |
Zyxin Protein | |
Research Use Only | |
ZYX | |
Recombinant Protein | |
Mammal | |
AAPRPSPAIS VSVSAPAFYA PQKKFGPVVA PKPKVNPFRP GDSEPPPAPG AQRAQMGRVG EIPPPPPEDF PLPPPPLAGD GDDAEGALGG AFPPPPPPIE ESFPPAPLEE EIFPSPPPPP EEEGGPEAPI PPPPQPREKV SSIDLEIDSL SSLLDDMTKN DPFKARVSSG YVPPPVATPF SSKSSTKPAA GGTAPLPPWK SPSSSQPLPQ VPAPAQSQTQ FHVQPQPQPK PQVQLHVQSQ TQPVSLANTQ PRGPPASSPA PAPKFSPVTP KFTPVASKFS PGAPGGSGSQ PNQKLGHPEA LSAGTGSPQP PSFTYAQQRE KPRVQEKQHP VPPPAQNQNQ VRSPGAPGPL TLKEVEELEQ LTQQLMQDME HPQRQNVAVN ELCGRCHQPL ARAQPAVRAL GQLFHIACFT CHQCAQQLQG QQFYSLEGAP YCEGCYTDTL EKCNTCGEPI TDRMLRATGK AYHPHCFTCV VCARPLEGTS FIVDQANRPH CVPDYHKQYA PRCSVCSEPI MPEPGRDETV RVVALDKNFH MKCYKCEDCG KPLSIEADDN GCFPLDGHVL CRKCHTARAQ T | |
Greater than 90% as determined by SDS-PAGE. |
7791 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |