missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ZSCAN21 Protein

A DNA sequence encoding the Homo sapiens (Human) ZSCAN21, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8774-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16056411

  • 160.00€ / 10 microgramos


Fecha estimada de envío
21-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_666019.1
ZSCAN21 Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
7589
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ZSCAN21
Recombinant Protein
E. coli
MMTKVLGMAP VLGPRPPQEQ VGPLMVKVEE KEEKGKYLPS LEMFRQRFRQ FGYHDTPGPR EALSQLRVLC CEWLRPEIHT KEQILELLVL EQFLTILPQE LQAWVQEHCP ESAEEAVTLL EDLERELDEP GHQVSTPPNE QKPVWEKISS SGTAKESPSS MQPQPLETSH KYESWGPLYI QESGEEQEFA QDPRKVRDCR LSTQHEESAD EQKGSEAEGL KGDIISVIIA NKPEASLERQ CVNLENEKGT KPPLQEAGSK KGRESVPTKP TPGERRYICA ECGKAFSNSS NLTKHRRTHT GEKPYVCTKC GKAFSHSSNL TLHYRTHLVD RPYDCKCGKA FGQSSDLLKH QRMHTEEAPY QCKDCGKAFS GKGSLIRHYR IHTGEKPYQC NECGKSFSQH AGLSSHQRLH TGEKPYKCKE CGKAFNHSSN FNKHHRIHTG EKPYWCHHCG KTFCSKSNLS K
Videos
Documentos