missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZSCAN20 Protein
A DNA sequence encoding the Homo sapiens (Human) ZSCAN20, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6913-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_660281.2 | |
ZSCAN20 Protein | |
Research Use Only | |
ZSCAN20 | |
Recombinant Protein | |
E. coli | |
MAMALELQAQ ASPQPEPEEL LIVKLEEDSW GSESKLWEKD RGSVSGPEAS RQRFRQFQYR DAAGPHEAFS QLWALCCRWL RPEIRLKEQI LELLVLEQFL TILPREVQTW VQARHPESGE EAVALVEDWH RETRTAGQSG LELHTEETRP LKTGEEAQSF QLQPVDPWPE GQSQKKGVKN TCPDLPNHLN AEVAPQPLKE SGVPVSKPSN TSEKEQGPEF WGLSLINSGK RSTADYSLDN EPAQALTWRD SRAWEEQYQW DVEDMKVSGV HWGYEETKTF LAILSESPFS EKLRTCHQNR QVYRAIAEQL RARGFLRTLE QCRYRVKNLL RNYRKAKSSH PPGTCPFYEE LEALVRARTA IRATDGPGEA VALPRLGYSD AEMDEQEEGG WDPEEMAEDC NGAGLVNVES TQGPRIAGAP ALFQSRIGKN MGV | |
Greater than 90% as determined by SDS-PAGE. |
7579 | |
500 μg | |
Not determined. | |
Full Length of Isoform 4 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |