missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zona pellucida sperm-binding protein 1

A DNA sequence encoding the Homo sapiens (Human) Zona pellucida sperm-binding protein 3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8806-ye-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16056581

  • 864.00€ / 200 microgramos


Fecha estimada de envío
21-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001103824.1
Zona pellucida sperm-binding protein 1
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
7784
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ZP3
Recombinant Protein
Yeast
QPLWLLQGGA SHPETSVQPV LVECQEATLM VMVSKDLFGT GKLIRAADLT LGPEACEPLV SMDTEDVVRF EVGLHECGNS MQVTDDALVY STFLLHDPRP VGNLSIVRTN RAEIPIECRY PRQGNVSSQA ILPTWLPFRT TVFSEEKLTF SLRLMEENWN AEKRSPTFHL GDAAHLQAEI HTGSHVPLRL FVDHCVATPT PDQNASPYHT IVDFHGCLVD GLTDASSAFK VPRPGPDTLQ FTVDVFHFAN DSRNMIYITC HLKVTLAEQD PDELNKACSF SKPSNSWFPV EGSADICQCC NKGDCGTPSH SRRQPHVMSQ WSRS
Videos
Documentos