missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zinc finger protein GLI2 Protein

A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein GLI2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6088-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919879

  • 726.00€ / 200 microgramos


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_005261.2
Zinc finger protein GLI2 Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
2736
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GLI2
Recombinant Protein
E. coli
EQLADLKEDL DRDDCKQEAE VVIYETNCHW EDCTKEYDTQ EQLVHHINNE HIHGEKKEFV CRWQACTREQ KPFKAQYMLV VHMRRHTGEK PHKCTFEGCS KAYSRLENLK THLRSHTGEK PYVCEHEGCN KAFSNASDRA KHQNRTHSNE KPYICKIPGC TKRYTDPSSL RKHVKTVHGP DAHVTKKQRN DVHLRTPLLK ENGDSEAGTE PGGPESTEAS STSQAVEDCL
Videos