missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZADH2 Protein
A cDNA sequence encoding the ZADH2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13989-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
284273 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMQKLVVT RLSPNFREAV TLSRDCPVPL PGDGDLLVRN RFVGVNASDI NYSAGRYDPS VKPPFDIGFE GIGEVVALGL SASARYTVGQ AVAYMAPGSF AEYTVVPASI ATPVPSVKPE YLTLLVSGTT AYISLKELGG LSEGKKVLVT AAAGGTGQFA MQLSKKAKCH VIGTCSSDEK SAFLKSLGCD RPINYKTEPV GTVLKQEYPE GVDVVYESVG GAMFDLAVDA LATKGRLIVI GFISGYQTPT GLSPVKAGTL PAKLLKKSAS VQGFFLNHYL SKYQAAMSHL LEMCVSGDLV CEVDLGDLSP EGRFTGLESI FRAVNYMYMG KNTGKIVVEL PHSVNSKL | |
Greater than 90.0% as determined by SDS-PAGE. |
ZADH2 Protein | |
Research Use Only | |
ZADH2 | |
Human | |
His | |
The ZADH2 solution (0.25 mg/1ml) contains Phosphate buffer saline (pH 7.4) and 20% glycerol. |