missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human XCL1 Protein
A DNA sequence encoding the Human XCL1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10221-ec-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002986.1 | |
XCL1 Protein | |
Research Use Only | |
XCL1 | |
The biological activity determined by a chemotaxis bioassay using human T-lymphocytes is in a concentration of 10-100 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
6375 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GSEVSDKRTC VSLTTQRLPV SRIKTYTITE GSLRAVIFIT KRGLKVCADP QATWVRDVVR SMDRKSNTRN NMIQTKPTGT QQSTNTAVTL TG | |
Greater than 98% as determined by SDS-PAGE and HPLC. |